Assay Details
Gene Symbol: RAB11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023655053.1 | XP_023510821.1 | 367 | 71 | ||
GenBank mRNA | JL621469.1 | - | 2 - 3 | 307 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
