Assay Details
Gene Symbol: GTF2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003363670.4 | XP_003363718.1 | 378 | 77 | ||
GenBank mRNA | JL616445.1 | - | 3 - 4 | 370 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
