Assay Details
Gene Symbol: AQP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500349.4 | XP_001500399.1 | 738 | 63 | ||
XM_005603021.3 | XP_005603078.1 | 743 | 63 | |||
XM_023616714.1 | XP_023472482.1 | 740 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
