Assay Details
Gene Symbol: ARPP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603042.3 | XP_005603099.2 | 244 | 113 | ||
XM_014733854.2 | XP_014589340.1 | 133 | 113 | |||
XM_023617235.1 | XP_023473003.1 | 266 | 113 | |||
GenBank mRNA | HQ890200.1 | - | 1 - 2 | 213 | 113 | |
JL615857.2 | - | 2 - 3 | 181 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
