Assay Details
Gene Symbol: MAPK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501448.6 | XP_001501498.1 | 2851 | 78 | ||
XM_023617312.1 | XP_023473080.1 | 1871 | 78 | |||
XM_023617320.1 | XP_023473088.1 | 1862 | 78 | |||
XM_023617327.1 | XP_023473095.1 | 1885 | 78 | |||
XM_023617337.1 | XP_023473105.1 | 1879 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
