Assay Details
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603057.3 | XP_005603114.1 | 845 | 95 | ||
XM_005603059.3 | XP_005603116.1 | 1369 | 95 | |||
XM_023617780.1 | XP_023473548.1 | 2771 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
