Assay Details
Gene Symbol: GATM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618259.1 | XP_023474027.1 | 1076 | 64 | ||
XM_023618268.1 | XP_023474036.1 | 1559 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
