Assay Details
Gene Symbol: SHF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603349.3 | XP_005603406.2 | 653 | 60 | ||
XM_014733935.2 | XP_014589421.2 | 653 | 60 | |||
XM_014733936.2 | XP_014589422.1 | 283 | 60 | |||
XM_014733937.2 | XP_014589423.2 | 653 | 60 | |||
XM_014733938.2 | XP_014589424.2 | 653 | 60 | |||
XM_023618367.1 | XP_023474135.1 | 653 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
