Assay Details
Gene Symbol: SORD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001918205.3 | XP_001918240.1 | 915 | 69 | ||
XM_023618424.1 | XP_023474192.1 | 1015 | 69 | |||
XM_023618426.1 | XP_023474194.1 | 781 | 69 | |||
XM_023618427.1 | XP_023474195.1 | 778 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
