Assay Details
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618492.1 | XP_023474260.1 | 2468 | 123 | ||
XM_023618497.1 | XP_023474265.1 | 3002 | 123 | |||
XM_023618503.1 | XP_023474271.1 | 2210 | 123 |

More Information
Set Membership: |
![]() ![]() |
