Assay Details
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618492.1 | XP_023474260.1 | 697 | 107 | ||
XM_023618503.1 | XP_023474271.1 | 439 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023618503.1 transcripts.
Set Membership: |
![]() ![]() |
