Assay Details
Gene Symbol: PLA2G4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734077.1 | XP_014589563.1 | 1148 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
