Assay Details
Gene Symbol: NDUFAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503436.5 | XP_001503486.1 | 1031 | 57 | ||
XM_005603128.3 | XP_005603185.1 | 1054 | 57 | |||
XM_023619619.1 | XP_023475387.1 | 849 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
