Assay Details
Gene Symbol: KNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734033.2 | XP_014589519.1 | 6120 | 63 | ||
XM_023619882.1 | XP_023475650.1 | 6165 | 63 | |||
XM_023619890.1 | XP_023475658.1 | 6063 | 63 | |||
XM_023619892.1 | XP_023475660.1 | 6114 | 63 | |||
XM_023619900.1 | XP_023475668.1 | 6121 | 63 | |||
XM_023619904.1 | XP_023475672.1 | 5794 | 63 | |||
XM_023619909.1 | XP_023475677.1 | 5788 | 63 | |||
XM_023619916.1 | XP_023475684.1 | 5760 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
