Assay Details
Gene Symbol: BAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619981.1 | XP_023475749.1 | 2005 | 57 | ||
XM_023619991.1 | XP_023475759.1 | 1846 | 57 | |||
XM_023619998.1 | XP_023475766.1 | 2005 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
