Assay Details
Gene Symbol: APEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001505131.5 | XP_001505181.4 | 673 | 58 | ||
XM_005603181.3 | XP_005603238.1 | 653 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
