Assay Details
Gene Symbol: TOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001505162.5 | XP_001505212.1 | 214 | 87 | ||
XM_023621446.1 | XP_023477214.1 | 214 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
