Assay Details
Gene Symbol: AP4S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603417.3 | XP_005603474.2 | 432 | 101 | ||
XM_014734145.2 | XP_014589631.2 | 658 | 101 | |||
GenBank mRNA | JL624871.1 | - | 1 - 2 | 84 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
