Assay Details
Gene Symbol: NUBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490020.5 | XP_001490070.1 | 487 | 73 | ||
XM_023623587.1 | XP_023479355.1 | 487 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
