Assay Details
Gene Symbol: ARHGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623610.1 | XP_023479378.1 | 4395 | 94 | ||
XM_023623619.1 | XP_023479387.1 | 4612 | 94 | |||
XM_023623630.1 | XP_023479398.1 | 4180 | 94 | |||
XM_023623636.1 | XP_023479404.1 | 6049 | 94 | |||
XM_023623643.1 | XP_023479411.1 | 4148 | 94 | |||
XM_023623648.1 | XP_023479416.1 | 4392 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
