Assay Details
Gene Symbol: TOGARAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624493.1 | XP_023480261.1 | 3424 | 87 | ||
XM_023624500.1 | XP_023480268.1 | 3423 | 87 | |||
XM_023624504.1 | XP_023480272.1 | 3426 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
