Assay Details
Gene Symbol: VCPKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624871.1 | XP_023480639.1 | 342 | 93 | ||
XM_023624878.1 | XP_023480646.1 | 354 | 93 | |||
XM_023624885.1 | XP_023480653.1 | 346 | 93 | |||
XM_023624887.1 | XP_023480655.1 | 740 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
