Assay Details
Gene Symbol: LRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649496.1 | XP_023505264.1 | 304 | 79 | ||
XM_023649497.1 | XP_023505265.1 | 306 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
