Assay Details
Gene Symbol: GARRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005596128.3 | XP_005596185.1 | 2996 | 60 | ||
XM_005596130.3 | XP_005596187.1 | 5052 | 60 | |||
XM_014733607.2 | XP_014589093.1 | 4591 | 60 | |||
XM_014733613.2 | XP_014589099.1 | 5052 | 60 | |||
XM_014733638.2 | XP_014589124.1 | 5052 | 60 | |||
XM_023649503.1 | XP_023505271.1 | 1554 | 60 | |||
XM_023649504.1 | XP_023505272.1 | 1539 | 60 | |||
XM_023649505.1 | XP_023505273.1 | 1101 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
