Assay Details
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496841.4 | XP_001496891.1 | 872 | 66 | ||
XM_005596226.3 | XP_005596283.1 | 857 | 66 | |||
XM_005596227.3 | XP_005596284.1 | 844 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
