Assay Details
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649776.1 | XP_023505544.1 | 1519 | 77 | ||
XM_023649777.1 | XP_023505545.1 | 1547 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
