Assay Details
Gene Symbol: DMAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500372.4 | XP_001500422.1 | 59 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001500372.4 transcripts.
Set Membership: |
![]() ![]() |
