Assay Details
Gene Symbol: DMRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499585.5 | XP_001499635.2 | 361 | 61 | ||
XM_005596294.3 | XP_005596351.1 | 649 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
