Assay Details
Gene Symbol: LYPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005596295.3 | XP_005596352.1 | 1310 | 68 | ||
XM_005596296.3 | XP_005596353.1 | 1348 | 68 | |||
XM_023651444.1 | XP_023507212.1 | 482 | 68 | |||
XM_023651445.1 | XP_023507213.1 | 1214 | 68 | |||
XM_023651446.1 | XP_023507214.1 | 462 | 68 | |||
XM_023651447.1 | XP_023507215.1 | 450 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
