Assay Details
Gene Symbol: CEACAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014728328.2 | XP_014583814.2 | 494 | 58 | ||
XM_014728367.2 | XP_014583853.2 | 474 | 58 | |||
XM_014728402.2 | XP_014583888.2 | 482 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
