Assay Details
Gene Symbol: NECTIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502277.5 | XP_001502327.2 | 312 | 95 | ||
XM_003362328.4 | XP_003362376.1 | 311 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
