Assay Details
Gene Symbol: APOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502376.4 | XP_001502426.1 | 81 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001502376.4 transcripts.
Set Membership: |
![]() ![]() |
