Assay Details
Gene Symbol: RELB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649886.1 | XP_023505654.1 | 899 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
