Assay Details
Gene Symbol: DMWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649933.1 | XP_023505701.1 | 2028 | 70 | ||
XM_023649935.1 | XP_023505703.1 | 1768 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
