Assay Details
Gene Symbol: SAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649985.1 | XP_023505753.1 | 623 | 59 | ||
XM_023649986.1 | XP_023505754.1 | 622 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
