Assay Details
Gene Symbol: CA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650045.1 | XP_023505813.1 | 1037 | 83 | ||
XM_023650047.1 | XP_023505815.1 | 1039 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
