Assay Details
Gene Symbol: AKT1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493191.5 | XP_001493241.2 | 519 | 111 | ||
XM_005596413.3 | XP_005596470.2 | 935 | 111 | |||
XM_023650191.1 | XP_023505959.1 | 555 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
