Assay Details
Gene Symbol: RDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494922.4 | XP_001494972.1 | 779 | 67 | ||
XM_005596534.3 | XP_005596591.1 | 620 | 67 | |||
XM_014730786.2 | XP_014586272.1 | 574 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
