Assay Details
Gene Symbol: MZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023651532.1 | XP_023507300.1 | 1522 | 70 | ||
XM_023651533.1 | XP_023507301.1 | 1441 | 70 | |||
XM_023651534.1 | XP_023507302.1 | 1963 | 70 | |||
XM_023651535.1 | XP_023507303.1 | 2317 | 70 | |||
XM_023651536.1 | XP_023507304.1 | 1614 | 70 | |||
XM_023651537.1 | XP_023507305.1 | 1252 | 70 | |||
XM_023651538.1 | XP_023507306.1 | 905 | 70 | |||
XM_023651539.1 | XP_023507307.1 | 391 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
