Assay Details
Gene Symbol: FILIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503595.5 | XP_001503645.1 | 3876 | 59 | ||
XM_005596796.3 | XP_005596853.1 | 3875 | 59 | |||
XM_014733831.2 | XP_014589317.1 | 3876 | 59 | |||
XM_023650661.1 | XP_023506429.1 | 4076 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
