Assay Details
Gene Symbol: UBE3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650710.1 | XP_023506478.1 | 954 | 68 | ||
XM_023650711.1 | XP_023506479.1 | 868 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
