Assay Details
Gene Symbol: DOP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650712.1 | XP_023506480.1 | 3340 | 95 | ||
XM_023650713.1 | XP_023506481.1 | 3388 | 95 | |||
XM_023650715.1 | XP_023506483.1 | 3233 | 95 | |||
XM_023650716.1 | XP_023506484.1 | 3313 | 95 | |||
XM_023650717.1 | XP_023506485.1 | 3310 | 95 | |||
XM_023650718.1 | XP_023506486.1 | 3307 | 95 | |||
XM_023650719.1 | XP_023506487.1 | 3340 | 95 | |||
XM_023650720.1 | XP_023506488.1 | 3101 | 95 | |||
XM_023650721.1 | XP_023506489.1 | 3277 | 95 | |||
XM_023650722.1 | XP_023506490.1 | 3128 | 95 | |||
XM_023650723.1 | XP_023506491.1 | 3241 | 95 | |||
XM_023650724.1 | XP_023506492.1 | 3101 | 95 |
