Assay Details
Gene Symbol: MRAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503691.4 | XP_001503741.2 | 765 | 77 | ||
XM_014734051.2 | XP_014589537.1 | 853 | 77 | |||
XM_023650754.1 | XP_023506522.1 | 441 | 77 | |||
XM_023650756.1 | XP_023506524.1 | 241 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
