Assay Details
Gene Symbol: FHL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005596838.3 | XP_005596895.1 | 515 | 66 | ||
XM_023650823.1 | XP_023506591.1 | 4417 | 66 | |||
XM_023650825.1 | XP_023506593.1 | 4481 | 66 | |||
XM_023650826.1 | XP_023506594.1 | 4273 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
