Assay Details
Gene Symbol: FAXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915544.5 | XP_001915579.2 | 1037 | 89 | ||
XM_023650842.1 | XP_023506610.1 | 943 | 89 | |||
XM_023650843.1 | XP_023506611.1 | 907 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
