Assay Details
Gene Symbol: COQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503863.4 | XP_001503913.1 | 915 | 65 | ||
XM_005596842.3 | XP_005596899.1 | 1016 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
