Assay Details
Gene Symbol: CRYBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650869.1 | XP_023506637.1 | 5542 | 100 | ||
GenBank mRNA | JL638035.1 | - | 4 - 5 | 465 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
