Assay Details
Gene Symbol: CRYBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650869.1 | XP_023506637.1 | 5576 | 66 | ||
GenBank mRNA | JL638035.1 | - | 5 - 6 | 499 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
