Assay Details
Gene Symbol: CRYBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650869.1 | XP_023506637.1 | 5958 | 81 | ||
GenBank mRNA | JL638035.1 | - | 8 - 9 | 881 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
