Assay Details
Gene Symbol: SCML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005596858.3 | XP_005596915.1 | 874 | 65 | ||
XM_023650875.1 | XP_023506643.1 | 878 | 65 | |||
XM_023650876.1 | XP_023506644.1 | 878 | 65 | |||
XM_023650877.1 | XP_023506645.1 | 806 | 65 | |||
XM_023650878.1 | XP_023506646.1 | 589 | 65 | |||
XM_023650879.1 | XP_023506647.1 | 400 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
